siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 17, 18 | 2169 | ||
XM_006234627.2 | XP_006234689.1 | 14, 15 | 1942 | |||
XM_006234629.2 | XP_006234691.1 | 12, 13 | 1708 | |||
XM_006234631.2 | XP_006234693.1 | 7, 8 | 1040 | |||
XM_008762055.1 | XP_008760277.1 | 13, 14 | 1851 | |||
XM_008762056.1 | XP_008760278.1 | 12, 13 | 1711 | |||
XM_008762057.1 | XP_008760279.1 | 11, 12 | 1619 | |||
XM_008762058.1 | XP_008760280.1 | 10, 11 | 1415 | |||
XM_008762059.1 | XP_008760281.1 | 17, 18 | 2306 | |||
GenBank mRNA | AF065147.1 | - | 7, 8 | 833 | ||
AF295360.1 | - | 3, 4 | 266 | |||
AF295361.1 | - | 3, 4 | 263 | |||
AF295362.1 | - | 3, 4 | 266 | |||
AF295363.1 | - | 5, 6 | 469 | |||
BC061531.1 | - | 7, 8 | 925 | |||
BC085703.1 | - | 7, 8 | 951 | |||
BC107949.1 | - | 7, 8 | 906 | |||
BC127485.1 | - | 7, 8 | 904 | |||
FQ234376.1 | - | 7, 8 | 921 | |||
M61874.1 | - | 10, 11 | 1338 | |||
M61875.1 | - | 7, 8 | 921 | |||
U46957.1 | - | 7, 8 | 833 | |||
U52179.1 | - | 7, 8 | 832 | |||
U96138.1 | - | 17, 18 | 2169 |
