siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 14 | 1815 | ||
XM_006234627.2 | XP_006234689.1 | 11 | 1588 | |||
XM_006234629.2 | XP_006234691.1 | 9 | 1354 | |||
XM_008762056.1 | XP_008760278.1 | 9 | 1357 | |||
XM_008762059.1 | XP_008760281.1 | 14 | 1952 | |||
GenBank mRNA | U96138.1 | - | 14 | 1815 |
