siRNA Details
Gene Symbol: Dpysl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012933.1 | NP_037065.1 | 2, 3 | 235 | ||
GenBank mRNA | CB735399.1 | - | 2, 3 | 231 | ||
CB802587.1 | - | 2, 3 | 250 | |||
U52103.1 | - | 1, 2 | 98 |
