siRNA Details
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012938.1 | NP_037070.1 | 6 | 738 | ||
XM_006249748.2 | XP_006249810.1 | 6 | 943 | |||
XM_006249749.2 | XP_006249811.1 | 7 | 1001 | |||
GenBank mRNA | BC062002.1 | - | 6 | 748 | 5599861 | |
D38104.1 | - | 6 | 738 | |||
D45187.1 | - | 6 | 880 |
