siRNA Details
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012955.1 | NP_037087.1 | 10 | 1017 | ||
XM_006257215.2 | XP_006257277.1 | 10 | 1155 | |||
XM_006257216.2 | XP_006257278.1 | 10 | 1316 | |||
XM_006257217.2 | XP_006257279.1 | 9 | 1030 | |||
XM_006257218.1 | XP_006257280.1 | 8 | 682 | |||
XM_008773403.1 | XP_008771625.1 | 9 | 1371 | |||
GenBank mRNA | X90355.1 | - | 10 | 1017 |
