siRNA Details
Gene Symbol: Gabrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012957.2 | NP_037089.1 | 2 | 97 | ||
XM_006246127.2 | XP_006246189.1 | 3 | 558 | |||
XM_008767635.1 | XP_008765857.1 | 3 | 562 | |||
GenBank mRNA | AY574251.1 | - | 2 | 97 | ||
X15467.1 | - | 2 | 173 |
