siRNA Details
Gene Symbol: Nedd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012986.1 | NP_037118.1 | 8, 9 | 656 | ||
XM_008766214.1 | XP_008764436.1 | 3, 4 | 4067 | |||
GenBank mRNA | CK365888.1 | - | 8, 9 | 656 | ||
U50842.1 | - | 8, 9 | 582 |
