siRNA Details
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013000.2 | NP_037132.2 | 19 | 2356 | ||
XM_006245594.2 | XP_006245656.1 | 19 | 2569 | |||
XM_006245595.2 | XP_006245657.1 | 19 | 2569 | |||
XM_006245596.2 | XP_006245658.1 | 19 | 2569 | |||
XM_006245597.2 | XP_006245659.1 | 19 | 2569 | |||
XM_006245598.2 | XP_006245660.1 | 18 | 2253 | |||
XM_006245599.2 | XP_006245661.1 | 18 | 2253 | |||
XM_008767360.1 | XP_008765582.1 | 19 | 2569 | |||
GenBank mRNA | M25719.1 | - | 18 | 2157 | ||
M25732.1 | - | 19 | 2356 | |||
X59685.1 | - | 16 | 1830 | |||
X59686.1 | - | 15 | 1511 | |||
X59687.1 | - | 18 | 1998 | |||
X59688.1 | - | 18 | 1987 | |||
X59689.1 | - | 18 | 1862 |
