siRNA Details
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013000.2 | NP_037132.2 | 24, 25 | 2922 | ||
XM_006245594.2 | XP_006245656.1 | 24, 25 | 3132 | |||
XM_006245596.2 | XP_006245658.1 | 23, 24 | 2931 | |||
XM_006245598.2 | XP_006245660.1 | 23, 24 | 2819 | |||
GenBank mRNA | M25719.1 | - | 23, 24 | 2723 | ||
M25732.1 | - | 24, 25 | 2922 | |||
X59685.1 | - | 21, 22 | 2396 | |||
X59686.1 | - | 20, 21 | 2077 | |||
X59687.1 | - | 22, 23 | 2360 |
