siRNA Details
Gene Symbol: Prkcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022507.1 | NP_071952.1 | 17 | 1781 | ||
XM_006239535.2 | XP_006239597.1 | 14 | 4276 | |||
XM_006239536.2 | XP_006239598.1 | 14 | 3482 | |||
XM_008764337.1 | XP_008762559.1 | 17 | 1682 | |||
GenBank mRNA | J04532.1 | - | 17 | 1781 | ||
M18332.1 | - | 14 | 1334 | |||
U27191.1 | - | 14 | 1546 |
