siRNA Details
Gene Symbol: Prkcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022507.1 | NP_071952.1 | 16, 17 | 1715 | ||
GenBank mRNA | J04532.1 | - | 16, 17 | 1715 | ||
M18332.1 | - | 13, 14 | 1268 | |||
U27191.1 | - | 13, 14 | 1480 |
