siRNA Details
Gene Symbol: Rab3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013018.2 | NP_037150.2 | 5 | 671 | ||
GenBank mRNA | BC087580.1 | - | 5 | 671 | 7314616 | |
FQ214415.1 | - | 5 | 639 | |||
X06889.1 | - | 5 | 643 |
