siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013034.4 | NP_037166.2 | 4 | 890 | ||
XM_008767950.1 | XP_008766172.1 | 5 | 946 | |||
GenBank mRNA | M79450.1 | - | 6 | 1014 | ||
X63253.1 | - | 4 | 898 | |||
X63995.1 | - | 5 | 943 | |||
Y11024.1 | - | 4 | 783 |
