siRNA Details
Gene Symbol: Abcc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013040.2 | NP_037172.2 | 22 | 2740 | ||
GenBank mRNA | AB045281.1 | - | 21 | 2528 | ||
AF019628.1 | - | 22 | 2740 | |||
AF087838.1 | - | 22 | 2740 | |||
D83598.1 | - | 22 | 2740 |
