siRNA Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB100593.1 | - | 5 | 984 | ||
AB100594.1 | - | 5 | 984 | |||
BC061756.1 | - | 5 | 988 | 5600029 | ||
X05300.1 | - | 5 | 989 |

More Information
Other Species Targeted

