siRNA Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB100593.1 | - | 9 | 1418 | ||
AB100594.1 | - | 9 | 1418 | |||
BC061756.1 | - | 9 | 1422 | 5600029 | ||
FQ220145.1 | - | 2 | 107 | |||
FQ229660.1 | - | 3 | 226 | |||
X05300.1 | - | 9 | 1423 |

More Information
Other Species Targeted

