siRNA Details
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013070.1 | NP_037202.1 | 58 | 8612 | ||
XM_006227637.2 | XP_006227699.1 | 57 | 8787 | |||
XM_008758593.1 | XP_008756815.1 | 58 | 8711 | |||
XM_008758594.1 | XP_008756816.1 | 58 | 8608 | |||
XM_008758595.1 | XP_008756817.1 | 57 | 8787 | |||
XM_008758596.1 | XP_008756818.1 | 57 | 8780 | |||
XM_008758597.1 | XP_008756819.1 | 57 | 8786 | |||
XM_008758598.1 | XP_008756820.1 | 8 | 2066 | |||
XM_008758599.1 | XP_008756821.1 | 8 | 1547 | |||
GenBank mRNA | AJ002967.1 | - | 58 | 8612 |
