siRNA Details
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013070.1 | NP_037202.1 | 27 | 3831 | ||
XM_006227637.2 | XP_006227699.1 | 26 | 4006 | |||
XM_008758593.1 | XP_008756815.1 | 27 | 3930 | |||
XM_008758594.1 | XP_008756816.1 | 27 | 3827 | |||
XM_008758595.1 | XP_008756817.1 | 26 | 4006 | |||
XM_008758596.1 | XP_008756818.1 | 26 | 3999 | |||
XM_008758597.1 | XP_008756819.1 | 26 | 4005 | |||
GenBank mRNA | AJ002967.1 | - | 27 | 3831 |
