siRNA Details
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013077.1 | NP_037209.1 | 1 | 26 | ||
GenBank mRNA | AB011544.1 | - | 1 | 26 |

More Information
Other Species Targeted

Gene Ontology Categories:
