siRNA Details
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100922.1 | NP_001094392.1 | 10 | 1115 | ||
XM_006255219.1 | XP_006255281.1 | 10 | 1049 | |||
GenBank mRNA | BC129128.1 | - | 10 | 1115 | 7314651 | |
FQ213798.1 | - | 2 | 113 | |||
X57986.1 | - | 10 | 1025 |
