siRNA Details
Gene Symbol: Cd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013121.1 | NP_037253.1 | 2 | 167 | ||
XM_008767078.1 | XP_008765300.1 | 2 | 811 | |||
XM_008767079.1 | XP_008765301.1 | 3 | 254 | |||
GenBank mRNA | X55288.1 | - | 2 | 167 |
