siRNA Details
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013123.3 | NP_037255.3 | 8 | 981 | ||
XM_006244746.2 | XP_006244808.1 | 9 | 3090 | |||
XM_006244747.2 | XP_006244809.1 | 9 | 1347 | |||
XM_006244748.2 | XP_006244810.1 | 8 | 1251 | |||
XM_006244749.2 | XP_006244811.1 | 10 | 1170 | |||
XM_006244750.2 | XP_006244812.1 | 10 | 3162 | |||
XM_006244751.2 | XP_006244813.1 | 9 | 1081 | |||
XM_006244752.2 | XP_006244814.1 | 9 | 1323 | |||
XM_006244753.2 | XP_006244815.1 | 9 | 2442 | |||
XM_006244754.2 | XP_006244816.1 | 8 | 2994 | |||
XM_008766993.1 | XP_008765215.1 | 9 | 1098 | |||
GenBank mRNA | AB079118.1 | - | 6 | 675 | ||
AB112436.1 | - | 10 | 1115 | |||
AB112437.1 | - | 8 | 981 | |||
AB112438.1 | - | 9 | 1053 | |||
M95578.1 | - | 9 | 1302 |
