siRNA Details
Gene Symbol: Itpr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013138.1 | NP_037270.1 | 19, 20 | 2564 | ||
XM_008772710.1 | XP_008770932.1 | 20, 21 | 2628 | |||
GenBank mRNA | GQ233031.1 | - | 19, 20 | 2430 | ||
L06096.1 | - | 19, 20 | 2564 |
