siRNA Details
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013151.2 | NP_037283.2 | 14 | 1652 | ||
XM_006253347.2 | XP_006253409.1 | 14 | 1724 | |||
XM_008771348.1 | XP_008769570.1 | 15 | 1782 | |||
GenBank mRNA | BC061565.1 | - | 14 | 1652 | 6920438 | |
M23697.1 | - | 13 | 1576 |
