siRNA Details
Gene Symbol: Vldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013155.2 | NP_037287.2 | 11, 12 | 1879 | ||
XM_006231197.2 | XP_006231259.1 | 12, 13 | 2184 | |||
XM_006231198.2 | XP_006231260.1 | 12, 13 | 1913 | |||
XM_006231199.2 | XP_006231261.1 | 11, 12 | 1787 | |||
GenBank mRNA | L35767.1 | - | 11, 12 | 1879 |
