siRNA Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013174.2 | NP_037306.1 | 6 | 1394 | ||
XM_006240366.1 | XP_006240428.1 | 7 | 1738 | |||
GenBank mRNA | BC092195.1 | - | 6 | 1381 | 7313608 | |
U03491.1 | - | 6 | 1394 |
