siRNA Details
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270889.1 | NP_001257818.1 | 4 | 745 | ||
NM_019144.2 | NP_062017.2 | 4 | 687 | |||
XM_006242691.2 | XP_006242753.1 | 6 | 1393 | |||
XM_006242692.2 | XP_006242754.1 | 5 | 1338 | |||
XM_006242693.2 | XP_006242755.1 | 4 | 1204 | |||
XM_006242694.2 | XP_006242756.1 | 4 | 641 | |||
GenBank mRNA | BC078847.1 | - | 4 | 745 | 7110955 | |
M76110.1 | - | 4 | 685 |
