siRNA Details
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013188.1 | NP_037320.1 | 19, 20 | 2495 | ||
GenBank mRNA | BC166475.1 | - | 2451 | 9100353 | ||
CO400919.1 | - | 5, 6 | 620 | |||
L10668.1 | - | 19, 20 | 2395 |
