siRNA Details
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013188.1 | NP_037320.1 | 7, 8 | 963 | ||
GenBank mRNA | BC166475.1 | - | 919 | 9100353 | ||
L10668.1 | - | 7, 8 | 863 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013188.1 | NP_037320.1 | 7, 8 | 963 | ||
GenBank mRNA | BC166475.1 | - | 919 | 9100353 | ||
L10668.1 | - | 7, 8 | 863 |