siRNA Details
Gene Symbol: Pfkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013190.4 | NP_037322.1 | 22 | 2391 | ||
XM_008772798.1 | XP_008771020.1 | 23 | 2494 | |||
GenBank mRNA | BC061791.1 | - | 22 | 2401 | 5598160 | |
X58865.1 | - | 22 | 2391 |
