siRNA Details
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013193.1 | NP_037325.1 | 6 | 958 | ||
XM_006251188.2 | XP_006251250.1 | 6 | 1008 | |||
XM_006251191.2 | XP_006251253.1 | 6 | 1008 | |||
GenBank mRNA | U61772.1 | - | 6 | 552 |
