siRNA Details
Gene Symbol: Slc35a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147140.2 | NP_671481.2 | 2 | 818 | ||
XM_006254528.1 | XP_006254590.1 | 2 | 852 | |||
XM_006254529.2 | XP_006254591.1 | 4 | 684 | |||
XM_008772019.1 | XP_008770241.1 | 2 | 771 | |||
GenBank mRNA | AF406814.1 | - | 1 | 589 | ||
BC085695.1 | - | 2 | 818 | 7191954 | ||
BC086986.1 | - | 2 | 771 | 7192444 | ||
FQ224389.1 | - | 4 | 689 |
