siRNA Details
Gene Symbol: Sdr9c7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241515.2 | XP_006241577.1 | 5, 6 | 5457 | ||
XM_006241516.2 | XP_006241578.1 | 5, 6 | 3726 | |||
XM_008765432.1 | XP_008763654.1 | 4, 5 | 5829 | |||
XM_008765433.1 | XP_008763655.1 | 5, 6 | 4435 | |||
GenBank mRNA | AY044435.1 | - | 2, 3 | 428 |
