siRNA Details
Gene Symbol: Ffar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153304.2 | NP_695216.1 | 1 | 560 | ||
GenBank mRNA | AB095744.1 | - | 1 | 558 | ||
AF539810.1 | - | 1 | 562 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ffar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153304.2 | NP_695216.1 | 1 | 560 | ||
GenBank mRNA | AB095744.1 | - | 1 | 558 | ||
AF539810.1 | - | 1 | 562 |
Set Membership: |
![]() |