siRNA Details
Gene Symbol: Zscan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728485.4 | XP_002728531.1 | 3 | 2306 | ||
GenBank mRNA | BC161917.1 | - | 3 | 2021 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zscan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728485.4 | XP_002728531.1 | 3 | 2306 | ||
GenBank mRNA | BC161917.1 | - | 3 | 2021 |