siRNA Details
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153622.2 | NP_705886.1 | 4 | 475 | ||
XM_006253941.2 | XP_006254003.1 | 7 | 1128 | |||
XM_006253942.2 | XP_006254004.1 | 7 | 1130 | |||
XM_006253944.2 | XP_006254006.1 | 7 | 1117 | |||
XM_006253946.2 | XP_006254008.1 | 6 | 1004 | |||
XM_006253947.2 | XP_006254009.1 | 7 | 1134 | |||
XM_006253948.2 | XP_006254010.1 | 2 | 1005 | |||
XM_006253949.2 | XP_006254011.1 | 5 | 887 | |||
XM_008771623.1 | XP_008769845.1 | 8 | 1205 | |||
XM_008771624.1 | XP_008769846.1 | 8 | 1193 | |||
XM_008771625.1 | XP_008769847.1 | 6 | 963 | |||
GenBank mRNA | AB025223.1 | - | 4 | 499 | ||
AB025224.1 | - | 4 | 499 | |||
BC078710.1 | - | 4 | 393 | 7110091 |
