siRNA Details
Gene Symbol: Mrgprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153722.2 | NP_714944.1 | 3 | 466 | ||
GenBank mRNA | M35297.1 | - | 469 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
