siRNA Details
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130536.1 | NP_001124008.1 | 4 | 1485 | ||
XM_008765449.1 | XP_008763671.1 | 3 | 1965 | |||
XM_008765450.1 | XP_008763672.1 | 3 | 1548 | |||
GenBank mRNA | AF481948.1 | - | 1 | 16 |
