siRNA Details
Gene Symbol: Clec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767505.1 | XP_008765727.1 | 20 | 2444 | ||
XM_008767506.1 | XP_008765728.1 | 19 | 2438 | |||
XM_008767507.1 | XP_008765729.1 | 20 | 2396 | |||
XM_008767508.1 | XP_008765730.1 | 19 | 2390 | |||
XM_008767509.1 | XP_008765731.1 | 19 | 2390 |
