siRNA Details
Gene Symbol: LOC100158225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127606.1 | NP_001121078.1 | 10 | 1295 | ||
GenBank mRNA | BC166509.1 | - | 10 | 1295 | 7127250 |

More Information
Other Species Targeted

