siRNA Details
Gene Symbol: LOC102555298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246066.2 | XP_006246128.1 | 26 | 3514 | ||
XM_006246068.2 | XP_006246130.1 | 26 | 3514 | |||
XM_006246069.2 | XP_006246131.1 | 23 | 2755 | |||
XM_008767618.1 | XP_008765840.1 | 26 | 3568 |
