siRNA Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007607.1 | NP_001007608.1 | 5, 6 | 1107 | ||
XM_006245942.2 | XP_006246004.1 | 5, 6 | 1186 | |||
XM_008767559.1 | XP_008765781.1 | 5, 6 | 1126 | |||
XM_008767560.1 | XP_008765782.1 | 5, 6 | 1153 | |||
GenBank mRNA | BC081875.1 | - | 5, 6 | 1107 | 7107681 |
