siRNA Details
Gene Symbol: Sec24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105780.1 | NP_001099250.1 | 24 | 3952 | ||
XM_006246241.2 | XP_006246303.1 | 23 | 5051 | |||
XM_006246242.2 | XP_006246304.1 | 23 | 5048 | |||
XM_006246243.2 | XP_006246305.1 | 22 | 4880 |
