siRNA Details
Gene Symbol: Obscn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767886.1 | XP_008766108.1 | 112 | 26128 | ||
GenBank mRNA | FQ224413.1 | - | 4 | 1452 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Obscn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767886.1 | XP_008766108.1 | 112 | 26128 | ||
GenBank mRNA | FQ224413.1 | - | 4 | 1452 |