siRNA Details
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191642.1 | NP_001178571.1 | 3 | 2565 | ||
XM_006246453.2 | XP_006246515.1 | 4 | 3007 | |||
XM_006246455.2 | XP_006246517.1 | 3 | 2668 | |||
XM_006246457.2 | XP_006246519.1 | 3 | 2668 | |||
XM_006246458.2 | XP_006246520.1 | 3 | 2670 |
