siRNA Details
Gene Symbol: Flii | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008279.1 | NP_001008280.1 | 9 | 898 | ||
XM_006246468.2 | XP_006246530.1 | 9 | 949 | |||
GenBank mRNA | BC085829.1 | - | 9 | 898 | 7133081 |
