siRNA Details
Gene Symbol: Adprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009246.1 | NP_001009246.1 | 3 | 248 | ||
XM_006246603.2 | XP_006246665.1 | 2 | 208 | |||
XM_006246604.2 | XP_006246666.1 | 3 | 315 | |||
GenBank mRNA | BC088174.1 | - | 3 | 248 | 7323419 | |
EU037900.1 | - | 1 | 95 |
