siRNA Details
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105812.1 | NP_001099282.1 | 14 | 1710 | ||
XM_006246893.2 | XP_006246955.1 | 15 | 2000 | |||
XM_006246894.2 | XP_006246956.1 | 15 | 1776 | |||
XM_006246895.2 | XP_006246957.1 | 15 | 1749 | |||
XM_006246896.2 | XP_006246958.1 | 15 | 2116 | |||
XM_006246897.2 | XP_006246959.1 | 14 | 1985 |
