siRNA Details
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105812.1 | NP_001099282.1 | 3 | 328 | ||
XM_006246893.2 | XP_006246955.1 | 3 | 606 | |||
XM_006246894.2 | XP_006246956.1 | 3 | 382 | |||
XM_006246895.2 | XP_006246957.1 | 3 | 355 | |||
XM_006246896.2 | XP_006246958.1 | 3 | 722 | |||
XM_006246897.2 | XP_006246959.1 | 3 | 603 |
