siRNA Details
Gene Symbol: Vps53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105813.1 | NP_001099283.1 | 22 | 2574 | ||
XM_006246899.2 | XP_006246961.1 | 21 | 2493 | |||
GenBank mRNA | BC168148.1 | - | 21 | 2489 | 7443082 |
